Learn More
Lonza™ Human Umbilical Vein Endothelial Cells
Product Code. 11655420
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
Each
Product Code. Quantity Unit Size
11655420 500 mL Each
1 options
Product Code. 11655420 Supplier Lonza™ Supplier No. CC3118
  • ONLINE EXCLUSIVE PRICE
    4925.00 DKK
    4537.88 DKK / Pakke med 1
    Spare 387.12 DKK
    7% Slukket
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Specifications
Form Liquid
For Use With (Application) Optimized to grow human small airway epithelial cells (SAEC).
Product Line SAGM BulletKit
Product Type Small Airway Epithelial Cell Growth Medium
Quantity 500 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • SAGM Bullet Kit
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P370+P378-In case of fire: Use water spray, alcohol-resistant foam, drychemical or carbondioxide to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, gentamicinsulfate