missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human ULK2 Synthetic Peptide
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
ULK2 Synthetic Peptide, PEP-1586, from Invitrogen.
ULK2, also known as ATG1B, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK2 results in a severe defect in the autophagy pathway. ULK2 is highly conserved among eukaryotes and shows high homology with its related protein ULK1. Both ULK1 and ULK2 form a complex with ATG13 and FIP200 that mediates TOR signaling and is essential for autophagy. Like ULK1, ULK2 is also thought to be involved in early neuronal growth and differentiation.
Tekniske data
Tekniske data
| Gen-id (Entrez) | 9706 |
| Arter | Human |
| Oprensningsmetode | purified |
| Adgangsnummer | Q8IYT8 |
| Fælles navn | Human ULK2 |
| Koncentration | 200 μg/mL |
| Indhold og opbevaring | -20°C |
| Oversigt | Synthetic Peptide |
| Format | Liquid |
| Formulering | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion