missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UBOX5 Full-length ORF (NP_055763.1, 1 a.a. - 541 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00022888-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MVINLCLPQFRPRIHCNKISADGYEVENLISEDLTKRSHGFRTEYFIKPPVYVTVSFPFNVEICRINIDLTAGGGQNVTGLEMYTSASSSRVSWNTPQCRTLGPAEPSVPDKEAFTLVGKVLLKNQSQVVFSHRGFKARPPFGAMEATLPSPAVVAQELWNKGALSLSHVAHLRICITHVTGGGIPCIKRLEVWGQPAKTCSQEVIDSILLVTSENLPQDVALQAPALPMESDCDPGDQPESQQAPSSLQKLAEIIQDVPEEFLDPITLEIMPCPMLLPSGKVIDQSTLEKCNRSEATWGRVPSDPFTGVAFTPHSQPLPHPSLKARIDHFLLQHSIPGCHLLGRAQTALAVIPSSIVLPSQKRKIEQAEHVPDSNFGVNASCFSATSPLVLPTTSEHTAKKMKATNEPSLTHMDCSTGPLSHEQKLSQSLEIALASTLGSMPSFTARLTRGQLQHLGTRGSNTSWRPGTGSEQPGSILGPECASCKRVFSPYFKKEPVYQLPCGHLLCRPCLGEKQRSLPMTCTACQRPVASQDVLRVHFSpecifications
NP_055763.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
85.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0860/RNF37/UBCE7IP5/UIP5 | |
UBOX5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
22888 | |
UBOX5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVINLCLPQFRPRIHCNKISADGYEVENLISEDLTKRSHGFRTEYFIKPPVYVTVSFPFNVEICRINIDLTAGGGQNVTGLEMYTSASSSRVSWNTPQCRTLGPAEPSVPDKEAFTLVGKVLLKNQSQVVFSHRGFKARPPFGAMEATLPSPAVVAQELWNKGALSLSHVAHLRICITHVTGGGIPCIKRLEVWGQPAKTCSQEVIDSILLVTSENLPQDVALQAPALPMESDCDPGDQPESQQAPSSLQKLAEIIQDVPEEFLDPITLEIMPCPMLLPSGKVIDQSTLEKCNRSEATWGRVPSDPFTGVAFTPHSQPLPHPSLKARIDHFLLQHSIPGCHLLGRAQTALAVIPSSIVLPSQKRKIEQAEHVPDSNFGVNASCFSATSPLVLPTTSEHTAKKMKATNEPSLTHMDCSTGPLSHEQKLSQSLEIALASTLGSMPSFTARLTRGQLQHLGTRGSNTSWRPGTGSEQPGSILGPECASCKRVFSPYFKKEPVYQLPCGHLLCRPCLGEKQRSLPMTCTACQRPVASQDVLRVHF | |
RUO | |
UBOX5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |