missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TSFM Partial ORF (NP_005717.2, 161 a.a. - 260 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_005717.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10102 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085215
|
Abnova™
H00010102-Q02.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075215
|
Abnova™
H00010102-Q02.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 [PubMed 17033963]).[supplied by OMIM]
Sequence: KVQWLTPVNLALWEAEAGGSLEGFLNSSELSGLPAGPDREGSLKDQLALAIGKLGENMILKRAAWVKVPSGFYVGSYVHGAMQSPSLHKLVLGKYGALVISpecifications
NP_005717.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COXPD3/EF-TS/EF-Tsmt | |
TSFM | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10102 | |
TSFM (Human) Recombinant Protein (Q02) | |
KVQWLTPVNLALWEAEAGGSLEGFLNSSELSGLPAGPDREGSLKDQLALAIGKLGENMILKRAAWVKVPSGFYVGSYVHGAMQSPSLHKLVLGKYGALVI | |
RUO | |
TSFM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |