missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Human TSC22D3 Synthetic Peptide

Artikelnummer. 15205479
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
15205479 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15205479 Leverandør Invitrogen™ Leverandørnr. PEP1549
 Begrænset vare
Forventet forsendelse: 18-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Synthetic Peptide

This peptide corresponds to 16 amino acids near the amino terminus of human TSC22D3. PEP-1549 can be used as a blocking peptide with polyclonal antibody PA5-34506.

The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.
TRUSTED_SUSTAINABILITY

Tekniske data

Gen-id (Entrez) 1831
Arter Human
Oprensningsmetode purified
Adgangsnummer Q99576
Fælles navn GILZ
Koncentration 200 μg/mL
Indhold og opbevaring -20°C
Oversigt Synthetic Peptide
Format Liquid
Formulering PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
Til brug med (applikation) Blocking Assay, Control
Gene Alias delta sleep inducing peptide, immunoreactor; delta sleep-inducing peptide immunoreactor; DIP; Dsip1; DSIPI; DSIP-immunoreactive leucine zipper protein; DSIP-immunoreactive peptide; GILZ; glucocorticoid-induced leucine zipper protein; long glucocorticoid-induced leucine zipper protein; Protein DIP; Tilz3; TSC22 domain family 3; TSC22 domain family member 3; TSC22 domain family protein 3; TSC22 domain family, member 3; TSC-22 related protein; TSC22D3; TSC-22-like protein; TSC-22R; TSC22-related inducible leucine zipper 3; TSC-22-related protein; TSC22-related-inducible leucine zipper 3
Gen symbol TSC22D3
Lyofiliseret, frysetørret No
Konserveringsmiddel Sodium azide
Produkttype Peptide
Mængde 50 μg
Regulatorisk status RUO
Opbevaringsbuffer PBS
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.