missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TRUB1 Full-length ORF (NP_631908.1, 1 a.a. - 349 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_631908.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 142940 |
Molecular Weight (g/mol) | 63.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138293
|
Abnova™
H00142940-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16128293
|
Abnova™
H00142940-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 [PubMed 12736709]).[supplied by OMIM]
Sequence: MAASEAAVVSSPSLKTDTSPVLETAGTVAAMAATPSARAAAAVVAAAARTGSEARVSKAALATKLLSLSGVFAVHKPKGPTSAELLNRLKEKLLAEAGMPSPEWTKRKKQTLKIGHGGTLDSAARGVLVVGIGSGTKMLTSMLSGSKRYTAIGELGKATDTLDSTGRVTEEKPYDKITQEDIEGILQKFTGNIMQVPPLYSALKKDGQRLSTLMKRGEVVEAKPARPVTVYSISLQKFQPPFFTLDVECGGGFYIRSLVSDIGKELSSCANVLELTRTKQGPFTLEEHALPEDKWTIDDIAQSLEHCSSLFPAELALKKSKPESNEQVLSCEYITLNEPKREDDVIKTCSpecifications
NP_631908.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAASEAAVVSSPSLKTDTSPVLETAGTVAAMAATPSARAAAAVVAAAARTGSEARVSKAALATKLLSLSGVFAVHKPKGPTSAELLNRLKEKLLAEAGMPSPEWTKRKKQTLKIGHGGTLDSAARGVLVVGIGSGTKMLTSMLSGSKRYTAIGELGKATDTLDSTGRVTEEKPYDKITQEDIEGILQKFTGNIMQVPPLYSALKKDGQRLSTLMKRGEVVEAKPARPVTVYSISLQKFQPPFFTLDVECGGGFYIRSLVSDIGKELSSCANVLELTRTKQGPFTLEEHALPEDKWTIDDIAQSLEHCSSLFPAELALKKSKPESNEQVLSCEYITLNEPKREDDVIKTC | |
RUO | |
TRUB1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
142940 | |
TRUB1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PUS4 | |
TRUB1 | |
Recombinant | |
wheat germ expression system |