missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIM26 Partial ORF (NP_003440.1, 146 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007726-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16064965

  • 4040.00 DKK / 25µg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6. [provided by RefSeq]

Sequence: REKILNHLSTLRRDRDKIQGFQAKGEADILAALKKLQDQRQYIVAEFEQGHQFLREREEHLLEQLAKLEQELTEGREKFKSRGVGELARLALVIS
Specifications

Specifications

NP_003440.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
REKILNHLSTLRRDRDKIQGFQAKGEADILAALKKLQDQRQYIVAEFEQGHQFLREREEHLLEQLAKLEQELTEGREKFKSRGVGELARLALVIS
RUO
TRIM26
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7726
TRIM26 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AFP/RNF95/ZNF173
TRIM26
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers