Learn More
Abnova™ Human TRA2A Full-length ORF (AAH17094, 1 a.a. - 282 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029896-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the transformer 2 homolog family and encodes a protein with two RS domains and an RRM (RNA recognition motif) domain. This phosphorylated nuclear protein binds to specific RNA sequences and plays a role in the regulation of pre-mRNA splicing. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: MSDVEENNFEGRESRSQSKSPTGTPARVKSESRSGSRSPSRVSKHSESHSRSRSKSRSRSRRHSHRRYTRSRSHSHSHRRRSRSRSYTPEYRRRRSRSHSPMSNRRRHTGSRANPDPNTCLGVFGLSLYTTERDLREVFSRYGPLSGVNVVYDQRTGRSRGFAFVYFERIDDSKEAMERANGMELDGRRIRVDYSITKRAHTPTPGIYMGRPTHSGGGGGGGGGGGGGGGGRRRDSYYDRGYDRGYDRYEDYDYRYRRRSPSPYYSRYRSRSRSRSYSPRRYSpecifications
AAH17094 | |
Liquid | |
29896 | |
TRA2A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSDVEENNFEGRESRSQSKSPTGTPARVKSESRSGSRSPSRVSKHSESHSRSRSKSRSRSRRHSHRRYTRSRSHSHSHRRRSRSRSYTPEYRRRRSRSHSPMSNRRRHTGSRANPDPNTCLGVFGLSLYTTERDLREVFSRYGPLSGVNVVYDQRTGRSRGFAFVYFERIDDSKEAMERANGMELDGRRIRVDYSITKRAHTPTPGIYMGRPTHSGGGGGGGGGGGGGGGGRRRDSYYDRGYDRGYDRYEDYDYRYRRRSPSPYYSRYRSRSRSRSYSPRRY | |
RUO | |
TRA2A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56.76kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSU53209 | |
TRA2A | |
Yes | |
wheat germ expression system |