missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TMOD2 Full-length ORF (AAH64961.1, 1 a.a. - 351 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00029767-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179082

  • 4040.00 DKK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a neuronal-specific member of the tropomodulin family of actin-regulatory proteins. The encoded protein caps the pointed end of actin filaments preventing both elongation and depolymerization. The capping activity of this protein is dependent on its association with tropomyosin. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: MALPFQKELEKYKNIDEDELLGKLSEEELKQLENVLDDLDPESAMLPAGFRQKDQTQKAATGPFDREHLLMYLEKEALEQKDREDFVPFTGEKKGRVFIPKEKPIETRKEEKVTLDPELEEALASASDTELYDLAAVLGVHNLLNNPKFDEETANNKGGKGPVRNVVKGEKVKPVFEEPPNPTNVEISLQQMKANDPSLQEVNLNNIKNIPIPTLREFAKALETNTHVKKFSLAATRSNDPVAIAFADMLKVNKTLTSLNIESNFITGTGILALVEALKENDTLTETKIDNQRQQLGTAVEMEIAQMLEENSRILKFGYQFTKQGPRTRVAAAITKNNDLVRKKRVEADRR
Specifications

Specifications

AAH64961.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.35kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC39481/N-TMOD/NTMOD
TMOD2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
29767
TMOD2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALPFQKELEKYKNIDEDELLGKLSEEELKQLENVLDDLDPESAMLPAGFRQKDQTQKAATGPFDREHLLMYLEKEALEQKDREDFVPFTGEKKGRVFIPKEKPIETRKEEKVTLDPELEEALASASDTELYDLAAVLGVHNLLNNPKFDEETANNKGGKGPVRNVVKGEKVKPVFEEPPNPTNVEISLQQMKANDPSLQEVNLNNIKNIPIPTLREFAKALETNTHVKKFSLAATRSNDPVAIAFADMLKVNKTLTSLNIESNFITGTGILALVEALKENDTLTETKIDNQRQQLGTAVEMEIAQMLEENSRILKFGYQFTKQGPRTRVAAAITKNNDLVRKKRVEADRR
RUO
TMOD2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers