Få mere at vide
Invitrogen™ Human TMEM51 Control Fragment Recombinant Protein
Beskrivelse
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-53190 (PA5-53190, PA5-64979 (PA5-64979. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Tekniske data
Tekniske data
| Adgangsnummer | Q9NW97 |
| Koncentration | ≥5.0 mg/mL |
| Til brug med (applikation) | Blocking Assay, Control |
| Formulering | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-id (Entrez) | 55092 |
| Navn | Human TMEM51 Control Fragment |
| Mængde | 100 μL |
| Regulatorisk status | RUO |
| Gene Alias | C1orf72; TMEM51; Transmembrane protein 51 |
| Fælles navn | TMEM51 |
| Vis mere |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.