missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TICAM2 Full-length ORF (NP_067681.1, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00353376-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180334

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TIRP is a Toll/interleukin-1 receptor (IL1R; MIM 147810) (TIR) domain-containing adaptor protein involved in Toll receptor signaling (see TLR4; MIM 603030).[supplied by OMIM]

Sequence: MGIGKSKINSCPLSLSWGKRHSVDTSPGYHESDSKKSEDLSLCNVAEHSNTTEGPTGKQEGAQSVEEMFEEEAEEEVFLKFVILHAEDDTDEALRVQNLLQDDFGIKPGIIFAEMPCGRQHLQNLDDAVNGSAWTILLLTENFLRDTWCNFQFYTSLMNSVNRQHKYNSVIPMRPLNNPLPRERTPFALQTINALEEESRGFPTQVERIFQESVYKTQQTIWKETRNMVQRQFIA
Specifications

Specifications

NP_067681.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC129876/MGC129877/TICAM-2/TIRAP3/TIRP/TRAM
TICAM2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
353376
TICAM2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGIGKSKINSCPLSLSWGKRHSVDTSPGYHESDSKKSEDLSLCNVAEHSNTTEGPTGKQEGAQSVEEMFEEEAEEEVFLKFVILHAEDDTDEALRVQNLLQDDFGIKPGIIFAEMPCGRQHLQNLDDAVNGSAWTILLLTENFLRDTWCNFQFYTSLMNSVNRQHKYNSVIPMRPLNNPLPRERTPFALQTINALEEESRGFPTQVERIFQESVYKTQQTIWKETRNMVQRQFIA
RUO
TICAM2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers