Learn More
Abnova™ Human TIAM2 Full-length ORF (NP_001010927.1, 1 a.a. - 626 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026230-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a guanine nucleotide exchange factor. A highly similar mouse protein specifically activates ras-related C3 botulinum substrate 1, converting this Rho-like guanosine triphosphatase (GTPase) from a guanosine diphosphate-bound inactive state to a guanosine triphosphate-bound active state. The encoded protein may play a role in neural cell development. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]
Sequence: MEGPRENQDPPPRPLARHLSDADRLRKVIQELVDTEKSYVKDLSCLFELYLEPLQNETFLTQDEMESLFGSLPEMLEFQKVFLETLEDGISASSDFNTLETPSQFRKLLFSLGGSFLYYADHFKLYSGFCANHIKVQKVLERAKTDKAFKAFLDARNPTKQHSSTLESYLIKPVQRVLKYPLLLKELVSLTDQESEEHYHLTEALKAMEKVASHINEMQKIYEDYGTVFDQLVAEQSGTEKEVTELSMGELLMHSTVSWLNPFLSLGKARKDLELTVFVFKRAVILVYKENCKLKKKLPSNSRPAHNSTDLDPFKFRWLIPISALQVRLGNPAGTENNSIWELIHTKSEIEGRPETIFQLCCSDSESKTNIVKVIRSILRENFRRHIKCELPLEKTCKDRLVPLKNRVPVSAKLASSRSLKVLKNSSSNEWTGETGKGTLLDSDEGSLSSGTQSSGCPTAEGRQDSKSTSPGKYPHPGLADFADNLIKESDILSDEDDDHRQTVKQGSPTKDIEIQFQRLRISEDPDVHPEAEQQPGPESGEGQKGGEQPKLVRGHFCPIKRKANSTKRDRGTLLKAQIRHQSLDSQSENATIDLNSVLEREFSVQSLTSVVSEECFYETESHGKSSpecifications
NP_001010927.1 | |
Liquid | |
26230 | |
TIAM2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEGPRENQDPPPRPLARHLSDADRLRKVIQELVDTEKSYVKDLSCLFELYLEPLQNETFLTQDEMESLFGSLPEMLEFQKVFLETLEDGISASSDFNTLETPSQFRKLLFSLGGSFLYYADHFKLYSGFCANHIKVQKVLERAKTDKAFKAFLDARNPTKQHSSTLESYLIKPVQRVLKYPLLLKELVSLTDQESEEHYHLTEALKAMEKVASHINEMQKIYEDYGTVFDQLVAEQSGTEKEVTELSMGELLMHSTVSWLNPFLSLGKARKDLELTVFVFKRAVILVYKENCKLKKKLPSNSRPAHNSTDLDPFKFRWLIPISALQVRLGNPAGTENNSIWELIHTKSEIEGRPETIFQLCCSDSESKTNIVKVIRSILRENFRRHIKCELPLEKTCKDRLVPLKNRVPVSAKLASSRSLKVLKNSSSNEWTGETGKGTLLDSDEGSLSSGTQSSGCPTAEGRQDSKSTSPGKYPHPGLADFADNLIKESDILSDEDDDHRQTVKQGSPTKDIEIQFQRLRISEDPDVHPEAEQQPGPESGEGQKGGEQPKLVRGHFCPIKRKANSTKRDRGTLLKAQIRHQSLDSQSENATIDLNSVLEREFSVQSLTSVVSEECFYETESHGKS | |
RUO | |
TIAM2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
97.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ41865/STEF | |
TIAM2 | |
Yes | |
wheat germ expression system |