missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human TBK1 Synthetic Peptide
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
NAK/TBK1 Synthetic Peptide, PEP-0093, from Invitrogen.
TBK1 serine/threonine kinase is a member of the IKK family and is involved in NF-kappa B activation and IRF3 signaling. TBK1 is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-KB mediates the expression of a great variety of genes in response to extracellular stimuli. NF-KB is associated with IKB proteins in the cell cytoplasm, which inhibit NF-KB activity. Phosphorylation of I-KB by IKB kinase (IKK) complex leads to degradation of I-KB and activation of NF-KB. The IKK complex contains IKKa, IKK beta, and IKKK. A novel IKK related kinase was recently identified and designated TBK1 (TANK-binding kinase 1), NAK (NF-KB-activating kinase), and T2K. NAK/TBK1 activates IKK beta through direct phosphorylation. NAK/TBK1 is activated by growth factors and PMA and mediates IKK and NF-kB activation in response to growth factors. NAK/TBK1 functions upstream of NIK and the IKK complex. NAK/TBK1 is also critical in protecting embryonic liver from apoptosis.
Tekniske data
Tekniske data
| Gen-id (Entrez) | 29110 |
| Arter | Human |
| Oprensningsmetode | purified |
| Adgangsnummer | Q9UHD2 |
| Fælles navn | Human TBK1 |
| Koncentration | 200 μg/mL |
| Indhold og opbevaring | -20°C |
| Oversigt | Synthetic Peptide |
| Format | Liquid |
| Formulering | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion