missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TBC1D7 Full-length ORF (NP_057579.1, 1 a.a. - 293 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

2665.00 DKK - 4040.00 DKK

Specifications

Accession Number NP_057579.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51256
Molecular Weight (g/mol) 60.4kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16179812
View Documents Promotion Details
Abnova™
H00051256-P01.25UG
25 ug
4040.00 DKK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16169812
View Documents Promotion Details
Abnova™
H00051256-P01.10UG
10 ug
2665.00 DKK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TBC1D7 belongs to the family of proteins sharing a 180- to 200-amino acid TBC domain presumed to have a role in regulating cell growth and differentiation. These proteins share significant homology with TRE2 (USP6; MIM 604334), yeast Bub2, and CDC16 (MIM 603461) (Nakashima et al., 2007 [PubMed 17658474]).[supplied by OMIM]

Sequence: MTEDSQRNFRSVYYEKVGFRGVEEKKSLEILLKDDRLDTEKLCTFSQRFPLPSMYRALVWKVLLGILPPHHESHAKVMMYRKEQYLDVLHALKVVRFVSDATPQAEVYLRMYQLESGKLPRSPSFPLEPDDEVFLAIAKAMEEMVEDSVDCYWITRRFVNQLNTKYRDSLPQLPKAFEQYLNLEDGRLLTHLRMCSAAPKLPYDLWFKRCFAGCLPESSLQRVWDKVVSGSCKILVFVAVEILLTFKIKVMALNSAEKITKFLENIPQDSSDAIVSKAIDLWHKHCGTPVHSS
Specifications

Specifications

NP_057579.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
60.4kDa
Glutathione Sepharose 4 Fast Flow
MTEDSQRNFRSVYYEKVGFRGVEEKKSLEILLKDDRLDTEKLCTFSQRFPLPSMYRALVWKVLLGILPPHHESHAKVMMYRKEQYLDVLHALKVVRFVSDATPQAEVYLRMYQLESGKLPRSPSFPLEPDDEVFLAIAKAMEEMVEDSVDCYWITRRFVNQLNTKYRDSLPQLPKAFEQYLNLEDGRLLTHLRMCSAAPKLPYDLWFKRCFAGCLPESSLQRVWDKVVSGSCKILVFVAVEILLTFKIKVMALNSAEKITKFLENIPQDSSDAIVSKAIDLWHKHCGTPVHSS
RUO
TBC1D7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
51256
TBC1D7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686N2317/FLJ32666/PIG51/TBC7/dJ257A7.3
TBC1D7
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications