missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TBC1D22B Full-length ORF (NP_060242.2, 1 a.a. - 505 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055633-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16131623

  • 4040.00 DKK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MAAENSKQFWKRSAKLPGSIQPVYGAQHPPLDPRLTKNFIKERSKVNTVPLKNKKASSFHEFARNTSDAWDIGDDEEEDFSSPSFQTLNSKVALATAAQVLENHSKLRVKPERSQSTTSDVPANYKVIKSSSDAQLSRNSSDTCLRNPLHKQQSLPLRPIIPLVARISDQNASGAPPMTVREKTRLEKFRQLLSSQNTDLDELRKCSWPGVPREVRPITWRLLSGYLPANTERRKLTLQRKREEYFGFIEQYYDSRNEEHHQDTYRQIHIDIPRTNPLIPLFQQPLVQEIFERILFIWAIRHPASGYVQGINDLVTPFFVVFLSEYVEEDVENFDVTNLSQDMLRSIEADSFWCMSKLLDGIQDNYTFAQPGIQKKVKALEELVSRIDEQVHNHFRRYEVEYLQFAFRWMNNLLMRELPLRCTIRLWDTYQSEPEGFSHFHLYVCAAFLIKWRKEILDEEDFQGLLMLLQNLPTIHWGNEEIGLLLAEAYRLKYMFADAPNHYRR
Specifications

Specifications

NP_060242.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
85.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf197/DKFZp762J0110/FLJ20337/MGC125626/MGC125627/RP4-744I24.2/dJ744I24.2
TBC1D22B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55633
TBC1D22B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAENSKQFWKRSAKLPGSIQPVYGAQHPPLDPRLTKNFIKERSKVNTVPLKNKKASSFHEFARNTSDAWDIGDDEEEDFSSPSFQTLNSKVALATAAQVLENHSKLRVKPERSQSTTSDVPANYKVIKSSSDAQLSRNSSDTCLRNPLHKQQSLPLRPIIPLVARISDQNASGAPPMTVREKTRLEKFRQLLSSQNTDLDELRKCSWPGVPREVRPITWRLLSGYLPANTERRKLTLQRKREEYFGFIEQYYDSRNEEHHQDTYRQIHIDIPRTNPLIPLFQQPLVQEIFERILFIWAIRHPASGYVQGINDLVTPFFVVFLSEYVEEDVENFDVTNLSQDMLRSIEADSFWCMSKLLDGIQDNYTFAQPGIQKKVKALEELVSRIDEQVHNHFRRYEVEYLQFAFRWMNNLLMRELPLRCTIRLWDTYQSEPEGFSHFHLYVCAAFLIKWRKEILDEEDFQGLLMLLQNLPTIHWGNEEIGLLLAEAYRLKYMFADAPNHYRR
RUO
TBC1D22B
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers