Learn More
Abnova™ Human TAF2 Partial ORF (AAC02966, 504 a.a. - 603 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006873-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the larger subunits of TFIID that is stably associated with the TFIID complex. It contributes to interactions at and downstream of the transcription initiation site, interactions that help determine transcription complex response to activators. [provided by RefSeq]
Sequence: LKSISNVSGKDIQPLIKQWVDQSGVVKFYGSFAFNRKRNVLELEIKQDYTSPGTQKYVGPLKVTVQELDGSFNHTLQIEENSLKHDIPCHSKSRRNKKKKSpecifications
AAC02966 | |
Liquid | |
6873 | |
TAF2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CIF150/TAF2B/TAFII150 | |
TAF2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LKSISNVSGKDIQPLIKQWVDQSGVVKFYGSFAFNRKRNVLELEIKQDYTSPGTQKYVGPLKVTVQELDGSFNHTLQIEENSLKHDIPCHSKSRRNKKKK | |
RUO | |
TAF2 | |
Wheat Germ (in vitro) | |
GST |