Learn More
Abnova™ Human SYT7 Partial ORF (NP_004191.2, 41 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009066-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Synaptotagmins, such as SYT7, are brain-specific calcium-dependent phospholipid-binding proteins that play a role in synaptic exocytosis and neurotransmitter release. See MIM 600782.[supplied by OMIM]
Sequence: CQRKLGKRYKNSLETVGTPDSGRGRSEKKAIKLPAGGKAVNTAPVPGQTPHDESDRRTEPRSSVSDLVNSLTSEMLMLSPGSEEDEAHEGCSRENLGRISpecifications
NP_004191.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CQRKLGKRYKNSLETVGTPDSGRGRSEKKAIKLPAGGKAVNTAPVPGQTPHDESDRRTEPRSSVSDLVNSLTSEMLMLSPGSEEDEAHEGCSRENLGRI | |
RUO | |
SYT7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9066 | |
SYT7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IPCA-7/MGC150517/PCANAP7/SYT-VII | |
SYT7 | |
Recombinant | |
wheat germ expression system |