Learn More
Abnova™ Human ST3GAL1 Full-length ORF (NP_003024.1, 1 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006482-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of the encoded protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B. Two transcript variants encoding the same protein have been found for this gene. Other transcript variants may exist, but have not been fully characterized yet. [provided by RefSeq]
Sequence: MVTLRKRTLKVLTFLVLFIFLTSFFLNYSHTMVATTWFPKQMVLELSENLKRLIKHRPCTCTHCIGQRKLSAWFDERFNQTMQPLLTAQNALLEDDTYRWWLRLQREKKPNNLNDTIKELFRVVPGNVDPMLEKRSVGCRRCAVVGNSGNLRESSYGPEIDSHDFVLRMNKAPTAGFEADVGTKTTHHLVYPESFRELGDNVSMILVPFKTIDLEWVVSAITTGTISHTYIPVPAKIRVKQDKILIYHPAFIKYVFDNWLQGHGRYPSTGILSVIFSMHVCDEVDLYGFGADSKGNWHHYWENNPSAGAFRKTGVHDADFESNVTATLASINKIRIFKGRSpecifications
NP_003024.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp666E036/DKFZp779K2051/FLJ36548/Gal-NAc6S/MGC9183/SIAT4A/SIATFL/ST3GalA/ST3GalA.1/ST3GalIA/ST3GalIA,1/ST3O | |
ST3GAL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6482 | |
ST3GAL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVTLRKRTLKVLTFLVLFIFLTSFFLNYSHTMVATTWFPKQMVLELSENLKRLIKHRPCTCTHCIGQRKLSAWFDERFNQTMQPLLTAQNALLEDDTYRWWLRLQREKKPNNLNDTIKELFRVVPGNVDPMLEKRSVGCRRCAVVGNSGNLRESSYGPEIDSHDFVLRMNKAPTAGFEADVGTKTTHHLVYPESFRELGDNVSMILVPFKTIDLEWVVSAITTGTISHTYIPVPAKIRVKQDKILIYHPAFIKYVFDNWLQGHGRYPSTGILSVIFSMHVCDEVDLYGFGADSKGNWHHYWENNPSAGAFRKTGVHDADFESNVTATLASINKIRIFKGR | |
RUO | |
ST3GAL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |