Learn More
Abnova™ Human SPAG16 Full-length ORF (NP_001020607.1, 1 a.a. - 183 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079582-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Cilia and flagella are comprised of a microtubular backbone, the axoneme, which is organized by the basal body and surrounded by plasma membrane. SPAG16 encodes 2 major proteins that associate with the axoneme of sperm tail and the nucleus of postmeiotic germ cells, respectively (Zhang et al., 2007 [PubMed 17699735]).[supplied by OMIM]
Sequence: MAAQRGMPSSAVRVLEEALGMGLTAAGDARDTADAVAAEGAYYLEQVTITEASEDDYEYEEIPDDNFSIPEGEEDLAKAIQMAQEQATDTEILERKTVLPSKHAVPEVIEDFLCNFLIKMGMTRTLDCFQSEWYELIQKGVTELRTVGNVPDVYTQIMLLENENKNLKKDLKHYKQAAEYVIFSpecifications
NP_001020607.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp666P1710/FLJ22724/FLJ37717/MGC87036/PF20/WDR29 | |
SPAG16 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79582 | |
SPAG16 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAQRGMPSSAVRVLEEALGMGLTAAGDARDTADAVAAEGAYYLEQVTITEASEDDYEYEEIPDDNFSIPEGEEDLAKAIQMAQEQATDTEILERKTVLPSKHAVPEVIEDFLCNFLIKMGMTRTLDCFQSEWYELIQKGVTELRTVGNVPDVYTQIMLLENENKNLKKDLKHYKQAAEYVIF | |
RUO | |
SPAG16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |