missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SORCS2 Partial ORF (NP_065828.1, 802 a.a. - 900 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_065828.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57537 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16179166
|
Abnova™
H00057537-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16169166
|
Abnova™
H00057537-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system. [provided by RefSeq]
Sequence: VLRVLDQFQVMPLQFSKELDAYNPNTPEWREDVGLVVTRLLSKETSVPQELLVTVVKPGLPTLADLYVLLPPPRPTRKRSLSSDKRLAAIQQVLNAQKISpecifications
NP_065828.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SORCS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57537 | |
SORCS2 (Human) Recombinant Protein (Q01) | |
VLRVLDQFQVMPLQFSKELDAYNPNTPEWREDVGLVVTRLLSKETSVPQELLVTVVKPGLPTLADLYVLLPPPRPTRKRSLSSDKRLAAIQQVLNAQKI | |
RUO | |
SORCS2 | |
Recombinant | |
wheat germ expression system |