missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Human Sonic Hedgehog C25II Recombinant Protein, PeproTech®
Klik for at se tilgængelige muligheder
Mængde:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
Beskrivelse
100-45-1MG will be provided as 2 x 500 μg (100-45-500UG). Recombinant E.coli-derived Human Sonic Hedgehog is a 20.0 kDa protein consisting of 176 amino acid residues, including an N-terminal Ile-Val-Ile sequence substituted for the naturally occurring, chemically modified, Cys residue. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Sonic Hedgehog (SHH), which is expressed only during embryogenesis, is instrumental in patterning the early embryo. It has been implicated as the key inductive signal in patterning of the ventral neural tube, the anterior-posterior limb axis, and the ventral somites. Of three human proteins showing sequence and functional similarity to the Sonic Hedgehog protein of Drosophila, this protein is the most similar. The protein is made as a precursor that is autocatalytically cleaved, the N-terminal portion is soluble and contains the signalling activity while the C-terminal portion is involved in precursor processing. More importantly, the C-terminal product covalently attaches a cholesterol moiety to the N-terminal product, restricting the N-terminal product to the cell surface and preventing it from freely diffusing throughout the developing embryo. Defects in this protein or in its signalling pathway are a cause of holoprosencephaly (HPE), a disorder in which the developing forebrain fails to correctly separate into right and left hemispheres. HPE is manifested by facial deformities. In addition, it is thought that mutations in this gene or in its signalling pathway may be responsible for VACTERL syndrome, which is characterized by vertebral defects, anal atresia, tracheoesophageal fistula with esophageal atresia, radial and renal dysplasia, cardiac anomalies, and limb abnormalities.
Tekniske data
Tekniske data
| Adgangsnummer | Q15465 |
| Til brug med (applikation) | Control, Bioactivity |
| Formulering | Protein with no preservative |
| Gen-id (Entrez) | 6469 |
| Molekylvægt (g/mol) | 20 kDa |
| Navn | Human Sonic Hedgehog C25II |
| Oprensningsmetode | Purified |
| Mængde | 100 μg |
| Kilde | E. coli |
| Opbevaringskrav | -20°C |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion