Learn More
Abnova™ Human SMARCA2 Partial ORF (NP_003061, 659 a.a. - 735 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_003061 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 6595 |
Molecular Weight (g/mol) | 34.21kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16145485
|
Abnova™
H00006595-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16135485
|
Abnova™
H00006595-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism. [provided by RefSeq]
Sequence: KILLDPNSEEVSEKDAKQIIETAKQDVDDEYSMQYSARGSQSYYTVAHAISERVEKQSALLINGTLKHYQLQGLEWMSpecifications
NP_003061 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.21kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAF190/BRM/FLJ36757/MGC74511/SNF2/SNF2L2/SNF2LA/SWI2/Sth1p/hBRM/hSNF2a | |
SMARCA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6595 | |
SMARCA2 (Human) Recombinant Protein (Q01) | |
KILLDPNSEEVSEKDAKQIIETAKQDVDDEYSMQYSARGSQSYYTVAHAISERVEKQSALLINGTLKHYQLQGLEWM | |
RUO | |
SMARCA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |