Learn More
Abcam Human SLC35A4 knockout HEK-293T cell line
Product Code. 32141153 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32141153 2 x 1,000,000 Cells Pack of 2
1 options
Product Code. 32141153 Supplier Abcam Supplier No. ab2665412x1000000Cells_
 Restricted Item
Forventet forsendelse: 07-12-2026
SLC35A4 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 3 and Insertion of the selection cassette in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial...
SLC35A4 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 3 and Insertion of the selection cassette in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.