missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC22A3 Partial ORF (NP_068812, 90 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006581-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135455

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq]

Sequence: CQRYLLEAANDSASATSALSCADPLAAFPNRSAPLVPCRGGWRYAQAHSTIVSEFDLVCVNAWMLD
Specifications

Specifications

NP_068812
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CQRYLLEAANDSASATSALSCADPLAAFPNRSAPLVPCRGGWRYAQAHSTIVSEFDLVCVNAWMLD
RUO
SLC22A3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6581
SLC22A3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EMT/EMTH/OCT3
SLC22A3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers