missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SIGLEC12 Partial ORF (NP_443729.1, 503 a.a. - 595 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00089858-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16065725

  • 2665.00 DKK / 10µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sialic acid-binding immunoglobulin-like lectins (SIGLECs) are a family of cell surface proteins belonging to the immunoglobulin superfamily. They mediate protein-carbohydrate interactions by selectively binding to different sialic acid moieties present on glycolipids and glycoproteins. This gene encodes a member of the SIGLEC3-like subfamily of SIGLECs. Members of this subfamily are characterized by an extracellular V-set immunoglobulin-like domain followed by two C2-set immunoglobulin-like domains, and the cytoplasmic tyrosine-based motifs ITIM and SLAM-like. The encoded protein, upon tyrosine phosphorylation, has been shown to recruit the Src homology 2 domain-containing protein-tyrosine phosphatases SHP1 and SHP2. It has been suggested that the protein is involved in the negative regulation of macrophage signaling by functioning as an inhibitory receptor. This gene is located in a cluster with other SIGLEC3-like genes on 19q13.4. Alternatively spliced transcript variants encoding distinct isoforms have been described for this gene. [provided by RefSeq]

Sequence: RSCRKKSARPAVGVGDTGMEDANAVRGSASQGPLIESPADDSPPHHAPPALATPSPEEGEIQYASLSFHKARPQYPQEQEAIGYEYSEINIPK
Specifications

Specifications

NP_443729.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.97kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RSCRKKSARPAVGVGDTGMEDANAVRGSASQGPLIESPADDSPPHHAPPALATPSPEEGEIQYASLSFHKARPQYPQEQEAIGYEYSEINIPK
RUO
SIGLEC12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
89858
SIGLEC12 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ38600/S2V/SIGLECL1/SLG/Siglec-12/Siglec-L1/Siglec-XII
SIGLEC12
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers