Learn More
Abnova™ Human SHANK2 Partial ORF (NP_036441, 1034 a.a. - 1124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_036441 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 22941 |
Molecular Weight (g/mol) | 35.75kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16134286
|
Abnova™
H00022941-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16124286
|
Abnova™
H00022941-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that is a member of the Shank family of synaptic proteins that may function as molecular scaffolds in the postsynaptic density (PSD). Shank proteins contain multiple domains for protein-protein interaction, including ankyrin repeats, an SH3 domain, a PSD-95/Dlg/ZO-1 domain, a sterile alpha motif domain, and a proline-rich region. This particular family member contains a PDZ domain, a consensus sequence for cortactin SH3 domain-binding peptides and a sterile alpha motif. The alternative splicing demonstrated in Shank genes has been suggested as a mechanism for regulating the molecular structure of Shank and the spectrum of Shank-interacting proteins in the PSDs of adult and developing brain. Two alternative splice variants, encoding distinct isoforms, are reported. Additional splice variants exist but their full-length nature has not been determined. [provided by RefSeq]
Sequence: SILQQMNREKLAKPGEGLDSPMGAKSASLAPRSPEIMSTISGTRSTTVTFTVRPGTSQPITLQSRPPDYESRTSGTRRAPSPVVSPTEMNKSpecifications
NP_036441 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CORTBP1/CTTNBP1/ProSAP1/SHANK/SPANK-3 | |
SHANK2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
22941 | |
SHANK2 (Human) Recombinant Protein (Q01) | |
SILQQMNREKLAKPGEGLDSPMGAKSASLAPRSPEIMSTISGTRSTTVTFTVRPGTSQPITLQSRPPDYESRTSGTRRAPSPVVSPTEMNK | |
RUO | |
SHANK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |