missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SCTR Full-length ORF (AAH35757.2, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006344-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180552

  • 4040.00 DKK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism. [provided by RefSeq]

Sequence: MRPHLSPPLQQLLLPVLLACAAHSTGALPRLCDVLQVLWEEQDQCLQELSREQTGDLGTEQPVPGCEGMWDNISCWPSSVPGRMVEVECPRFLRMLTSRNGSLFRNCTQDGWSETFPRPNLACGVNVNDSSNEKRHSYLLKLKVMYTVGYSSSLVMLLVALGILCAFRRLHCTRNYIHMHLFVSFILRALSNFIKDAVLFSSDDVTYCDAHRAGCKLIMVLFQYCIMANYSWLLVEGLYLHTLLAISFFSERKYLQGFVAFGWGSPAIFVALWAIARHFLEDVGCWDINANASIWWIIRGPVILSILINFILFINILRILMRKLRTQETRGNEVSHYKRLARSTLLLIPLFGIHYIVFAFSPEDAMEIQLFFELALGSFQGLVVAVLYCFLNGEVQLEVQKKWQQWHLREFPLHPVASFSNSTKASHLEQSQGTCRTSII
Specifications

Specifications

AAH35757.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
76.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SR
SCTR
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
6344
SCTR (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRPHLSPPLQQLLLPVLLACAAHSTGALPRLCDVLQVLWEEQDQCLQELSREQTGDLGTEQPVPGCEGMWDNISCWPSSVPGRMVEVECPRFLRMLTSRNGSLFRNCTQDGWSETFPRPNLACGVNVNDSSNEKRHSYLLKLKVMYTVGYSSSLVMLLVALGILCAFRRLHCTRNYIHMHLFVSFILRALSNFIKDAVLFSSDDVTYCDAHRAGCKLIMVLFQYCIMANYSWLLVEGLYLHTLLAISFFSERKYLQGFVAFGWGSPAIFVALWAIARHFLEDVGCWDINANASIWWIIRGPVILSILINFILFINILRILMRKLRTQETRGNEVSHYKRLARSTLLLIPLFGIHYIVFAFSPEDAMEIQLFFELALGSFQGLVVAVLYCFLNGEVQLEVQKKWQQWHLREFPLHPVASFSNSTKASHLEQSQGTCRTSII
RUO
SCTR
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers