Learn More
Abnova™ Human SCTR Full-length ORF (AAH35757.2, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006344-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism. [provided by RefSeq]
Sequence: MRPHLSPPLQQLLLPVLLACAAHSTGALPRLCDVLQVLWEEQDQCLQELSREQTGDLGTEQPVPGCEGMWDNISCWPSSVPGRMVEVECPRFLRMLTSRNGSLFRNCTQDGWSETFPRPNLACGVNVNDSSNEKRHSYLLKLKVMYTVGYSSSLVMLLVALGILCAFRRLHCTRNYIHMHLFVSFILRALSNFIKDAVLFSSDDVTYCDAHRAGCKLIMVLFQYCIMANYSWLLVEGLYLHTLLAISFFSERKYLQGFVAFGWGSPAIFVALWAIARHFLEDVGCWDINANASIWWIIRGPVILSILINFILFINILRILMRKLRTQETRGNEVSHYKRLARSTLLLIPLFGIHYIVFAFSPEDAMEIQLFFELALGSFQGLVVAVLYCFLNGEVQLEVQKKWQQWHLREFPLHPVASFSNSTKASHLEQSQGTCRTSIISpecifications
AAH35757.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SR | |
SCTR | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6344 | |
SCTR (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRPHLSPPLQQLLLPVLLACAAHSTGALPRLCDVLQVLWEEQDQCLQELSREQTGDLGTEQPVPGCEGMWDNISCWPSSVPGRMVEVECPRFLRMLTSRNGSLFRNCTQDGWSETFPRPNLACGVNVNDSSNEKRHSYLLKLKVMYTVGYSSSLVMLLVALGILCAFRRLHCTRNYIHMHLFVSFILRALSNFIKDAVLFSSDDVTYCDAHRAGCKLIMVLFQYCIMANYSWLLVEGLYLHTLLAISFFSERKYLQGFVAFGWGSPAIFVALWAIARHFLEDVGCWDINANASIWWIIRGPVILSILINFILFINILRILMRKLRTQETRGNEVSHYKRLARSTLLLIPLFGIHYIVFAFSPEDAMEIQLFFELALGSFQGLVVAVLYCFLNGEVQLEVQKKWQQWHLREFPLHPVASFSNSTKASHLEQSQGTCRTSII | |
RUO | |
SCTR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |