missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SAV1 Partial ORF (NP_068590, 300 a.a. - 383 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00060485-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179416

  • 4040.00 DKK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 2 WW domains and a coiled-coil region. It is ubiquitously expressed in adult tissues. The encoded protein is 94% identical to the mouse protein at the amino acid level. [provided by RefSeq]

Sequence: HTAEIPDWLQVYARAPVKYDHILKWELFQLADLDTYQGMLKLLFMKELEQIVKMYEAYRQALLTELENRKQRQQWYAQQHGKNF
Specifications

Specifications

NP_068590
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.98kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HTAEIPDWLQVYARAPVKYDHILKWELFQLADLDTYQGMLKLLFMKELEQIVKMYEAYRQALLTELENRKQRQQWYAQQHGKNF
RUO
SAV1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
60485
SAV1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SAV/WW45/WWP4
SAV1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers