missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA Kit

ELISA

5180.00 DKK - 36310.00 DKK

Specifications

Accession Number Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2
Assay Range 62.5 to 4000 Units/mL
Assay Sensitivity <62.5 U/mL
Conjugate HRP
Product Type ELISA
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17198865
View Documents Promotion Details
Invitrogen™
BMS2323
96 Tests
5180.00 DKK
Pack of 96
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17124769
View Documents
Invitrogen™
BMS2323TWO
2 x 96 Tests
8230.00 DKK
Pack of 2
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17114769
View Documents
Invitrogen™
BMS2323FIVE
5 x 96 Tests
18430.00 DKK
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17108875
View Documents
Invitrogen™
BMS2323TEN
10 x 96 Tests
36310.00 DKK
Pack of 10
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA is a serology assay that measures and quantitates immunoglobulin (Ig) antibodies against SARS-CoV-2 Spike (Trimer) in human serum or plasma. Principle of the method The Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of Ig antibodies bound to SARS-CoV-2 Spike (Trimer). A trimerized Spike protein is pre-coated in the wells of the supplied microplate. Samples, controls, including a high control that can be used as a standard are then added into these wells and bind to the immobilized (capture) Spike protein. The wells are washed and Ig antibodies conjugated to HRP are added and will bind to any captured antibodies. The Ig Total antibody recognizes IgG, IgM and IgA. The wells are washed and a substrate solution is added that reacts with the enzyme complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of antibody present in the original specimen.

It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
Specifications

Specifications

Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2
<62.5 U/mL
ELISA
Colorimetric Microplate Reader
4.3
HRP
Plasma 5 ÎĽL, Serum 5 ÎĽL
SARS-CoV-2 Spike (trimer) Protein
1 hrs 15 mins
62.5 to 4000 Units/mL
HRP
Plasma, Serum
5%
Pre-coated 96 well plate, High Control (standard), Calibrator (medium control), Low Control, Anti-Ig Total - HRP conjugate, Assay Buffer concentrate, Wash Buffer concentrate, Substrate solution, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
30 mins
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human SARS-CoV-2 Spike (Trimer) Ig Total ELISA Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.