Learn More
Invitrogen™ Human SAMD9L (aa 1245-1355) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP92806
Description
Highest antigen sequence indentity to the following orthologs: Mouse (67%), Rat (67%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53991 (PA5-53991. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The function of this protein has not been specifically defined.
Specifications
Q8IVG5 | |
Blocking Assay, Control | |
219285 | |
100 ÎĽL | |
AA175286; C7orf6; DRIF2; ESTM25; KIAA2005; mKIAA2005; SAM domain-containing protein 9-like; SAMD9L; sterile alpha motif domain containing 9 like; sterile alpha motif domain containing 9-like; sterile alpha motif domain-containing protein 9-like; UEF; UEF1; v-myc myelocytomatosis viral related oncogene, neuroblastoma derived | |
SAMD9L | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human SAMD9L (aa 1245-1355) Control Fragment | |
RUO | |
SAMD9L | |
Unconjugated | |
Recombinant | |
ECYLALSKFTSHLKNLQSDLKRCFDFFIDYMVLLKMRYTQKEIAEIMLSKKVSRCFRKYTELFCHLDPCLLQSKESQLLQEENCRKKLEALRADRFAGLLEYLNPNYKDAT | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.