missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SAGE1 Partial ORF (NP_061136, 357 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_061136 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55511 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148176
|
Abnova™
H00055511-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138176
|
Abnova™
H00055511-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas. [provided by RefSeq]
Sequence: PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDNSpecifications
NP_061136 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SAGE | |
SAGE1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55511 | |
SAGE1 (Human) Recombinant Protein (Q01) | |
PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDN | |
RUO | |
SAGE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |