Få mere at vide
R&D Systems™ Human S100B DuoSet ELISA
Mængde:
1 sæt (til 5 tallerkener)
Beskrivelse
Indeholder
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human S100B DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human S100B.
Tekniske data
Tekniske data
| Mængde | 1 sæt (til 5 tallerkener) |
| Arter | Human |
| Konjugeret | Biotin |
| Bindende mål | S100B |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 6285 |
| Synonym | beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta |
| Indhold og opbevaring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |