Learn More
Abnova™ Human RSU1 Partial ORF (NP_036557.1, 178 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006251-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is involved in the Ras signal transduction pathway, growth inhibition, and nerve-growth factor induced differentiation processes, as determined in mouse and human cell line studies. In mouse, the encoded protein was initially isolated based on its ability to inhibit v-Ras transformation. Multiple alternatively spliced transcript variants for this gene have been reported; one of these variants was found only in glioma tumors. [provided by RefSeq]
Sequence: ELTQLKELHIQGNRLTVLPPELGNLDLTGQKQVFKAENNPWVTPIADQFQLGVSHVFEYIRSETYKYLYGRHMQANPEPPKKNNDKSKKISRKPLAAKNRSpecifications
NP_036557.1 | |
Liquid | |
6251 | |
RSU1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31034/RSP-1 | |
RSU1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ELTQLKELHIQGNRLTVLPPELGNLDLTGQKQVFKAENNPWVTPIADQFQLGVSHVFEYIRSETYKYLYGRHMQANPEPPKKNNDKSKKISRKPLAAKNR | |
RUO | |
RSU1 | |
Wheat Germ (in vitro) | |
GST |