missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RPS28 Partial ORF (NP_001022, 1 a.a. - 55 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006234-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134765

  • 4150.00 DKK / 25µg
Estimated Shipment: 19-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S28E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. [provided by RefSeq]

Sequence: MDTSRVQPIKLARVTKVLGRTGSQGQCTQVRVEFMDDTSRSIIRNVKGPVREGDV
Specifications

Specifications

NP_001022
Liquid
6234
RPS28 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RPS28
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.79kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDTSRVQPIKLARVTKVLGRTGSQGQCTQVRVEFMDDTSRSIIRNVKGPVREGDV
RUO
RPS28
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers