Learn More
Abnova™ Human RP9 Partial ORF (NP_976033.1, 66 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006100-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene can be bound and phosphorylated by the protooncogene PIM1 product, a serine/threonine protein kinase . This protein localizes in nuclear speckles containing the splicing factors, and has a role in pre-mRNA splicing. CBF1-interacting protein (CIR), a corepressor of CBF1, can also bind to this protein and effects alternative splicing. Mutations in this gene result in autosomal dominant retinitis pigmentosa-9. This gene has a pseudogene (GeneID: 441212), which is located in tandem array approximately 166 kb distal to this gene. [provided by RefSeq]
Sequence: KPEDCIPDVPGNEHAREFLAHAPTKGLWMPLGKEVKVMQCWRCKRYGHRTGDKECPFFIKGNQKLEQFRVAHEDPMYDIIRDNKRHEKDVRIQQLKQLLESpecifications
NP_976033.1 | |
Liquid | |
6100 | |
RP9 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PAP-1 | |
RP9 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KPEDCIPDVPGNEHAREFLAHAPTKGLWMPLGKEVKVMQCWRCKRYGHRTGDKECPFFIKGNQKLEQFRVAHEDPMYDIIRDNKRHEKDVRIQQLKQLLE | |
RUO | |
RP9 | |
Wheat Germ (in vitro) | |
GST |