missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF123 Partial ORF (NP_071347, 1216 a.a. - 1314 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_071347 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 63891 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189466
|
Abnova™
H00063891-Q01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16179466
|
Abnova™
H00063891-Q01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. [provided by RefSeq]
Sequence: ADYISADELAQVEQMLAHLTSASAQAAAASLPTSEEDLCPICYAHPISAVFQPCGHKSCKACINQHLMNNKDCFFCKTTIVSVEDWEKGANTSTTSSAASpecifications
NP_071347 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686C2222/FLJ12565/FP1477/KPC1/MGC163504 | |
RNF123 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
63891 | |
RNF123 (Human) Recombinant Protein (Q01) | |
ADYISADELAQVEQMLAHLTSASAQAAAASLPTSEEDLCPICYAHPISAVFQPCGHKSCKACINQHLMNNKDCFFCKTTIVSVEDWEKGANTSTTSSAA | |
RUO | |
RNF123 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |