Learn More
Abnova™ Human RGS11 Partial ORF (NP_003825, 101 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008786-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the RGS (regulator of G protein signaling) family. Members of the RGS family act as GTPase-activating proteins on the alpha subunits of heterotrimeric, signal-transducing G proteins. This protein inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits, thereby driving them into their inactive GDP-bound form. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: DYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMTSpecifications
NP_003825 | |
Liquid | |
8786 | |
RGS11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RS11 | |
RGS11 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMT | |
RUO | |
RGS11 | |
Wheat Germ (in vitro) | |
GST |