Få mere at vide
R&D Systems™ Human Reg3A DuoSet ELISA
Artikelnummer. 30000986
Skift visning
Format:
96-brønds plader
Artikelnummer. Format
30000986 96-brønds plader
1 options
Artikelnummer. 30000986 Leverandør R&D Systems™ Leverandørnr. DY594005
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
For the determination of natural and recombinant Reg3A in humans. R&D Systems Human Reg3A DuoSet ELISA Kit is a solid phase sandwich ELISA suitable for the analysis of cell culture supernate, serum and plasma samples.
The Human Reg3A DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Reg3A.
TRUSTED_SUSTAINABILITY
Tekniske data
Produkttype ELISA Kit
Arter Human
Bindende mål Reg3A
Format 96-brønds plader
Til brug med (applikation) ELISA
Gen-id (Entrez) 5068
Prøvetype Cell Culture Supernatant, Serum, Plasma
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Research Use Only.