Få mere at vide
STEMCELL Technologies Human Recombinant bFGF
Mængde:
10 μg
1000 μg
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
- Basic fibroblast growth factor (bFGF) is a prototypic member of the fibroblast growth factor family
- Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman & Klagsbrun; Kimelman & Kirschner) and are involved in a variety of biological processes including cell proliferation, differentiation, survival, and apoptosis (Folkman & Klagsbrun; Klagsbrun; Rifkin & Moscatelli)
- bFGF has the β-trefoil structure (Ponting & Russell), binds to the four FGF receptor (FGFR) family members, and activates JAK/STAT, PI3K, ERK1/2, and other receptor tyrosine kinase (RTK) signaling pathways
- It supports the maintenance of undifferentiated human embryonic stem cells (Xu et al.; Kang et al.), stimulates human embryonic stem cells to form neural rosettes (Zhang et al.), and improves proliferation of human mesenchymal stem cells and enhances chondrogenic differentiation (Solchaga et al.)
Tekniske data
Tekniske data
| Navn | Basic fibroblast growth factor, FGF-β, FGF2, Fibroblast growth factor-basic, HBGF-2 |
| Mængde | 10 μg |
| Celletype | Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
| Renhed eller kvalitetsklasse | >95% |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.