Learn More
Abnova™ Human RDH8 Full-length ORF (AAI46480.1, 1 a.a. - 311 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | AAI46480.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 50700 |
Molecular Weight (g/mol) | 61.16kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16189412
|
Abnova™
H00050700-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16179412
|
Abnova™
H00050700-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
All-trans-retinol dehydrogenase (RDH8) is a visual cycle enzyme that reduces all-trans-retinal to all-trans-retinol in the presence of NADPH (Rattner et al., 2000 [PubMed 10753906]). It is a member of the short chain dehydrogenase/reductase family and is located in the outer segments of photoreceptors; hence it is also known as photoreceptor retinol dehydrogenase. It is important in the visual cycle by beginning the rhodopsin regeneration pathway by reducing all-trans-retinal, the product of bleached and hydrolysed rhodopsin (Rando, 2001 [PubMed 11710234]). This is a rate-limiting step in the visual cycle (Saari et al., 1998 [PubMed 9667000]).[supplied by OMIM]
Sequence: MAAAPRTVLISGCSSGIGLELAVQLAHDPKKRYQVVATMRDLGKKETLEAAAGEALGQTLTVAQLDVCSDESVAQCLSCIQGEVDVLVNNAGMGLVGPLEGLSLAAMQNVFDTNFFGAVRLVKAVLPGMKRRRQGHIVVISSVMGLQGVIFNDVYAASKFALEGFFESLAIQLLQFNIFISLVEPGPVVTEFEGKLLAQVSMAEFPGTDPETLHYFRDLYLPASRKLFCSVGQNPQDVVQAIVNVISSTRPPLRRQTNIRYSPLTTLKTVDSSGSLYVRTTHRLLFRCPRLLNLGLQCLSCGCLPTRVRPRSpecifications
AAI46480.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.16kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAAPRTVLISGCSSGIGLELAVQLAHDPKKRYQVVATMRDLGKKETLEAAAGEALGQTLTVAQLDVCSDESVAQCLSCIQGEVDVLVNNAGMGLVGPLEGLSLAAMQNVFDTNFFGAVRLVKAVLPGMKRRRQGHIVVISSVMGLQGVIFNDVYAASKFALEGFFESLAIQLLQFNIFISLVEPGPVVTEFEGKLLAQVSMAEFPGTDPETLHYFRDLYLPASRKLFCSVGQNPQDVVQAIVNVISSTRPPLRRQTNIRYSPLTTLKTVDSSGSLYVRTTHRLLFRCPRLLNLGLQCLSCGCLPTRVRPR | |
RUO | |
RDH8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
50700 | |
RDH8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PRRDH/SDR28C2 | |
RDH8 | |
Recombinant | |
wheat germ expression system |