missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RBM34 Full-length ORF (NP_055829.1, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Spécification
Accession Number | NP_055829.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23029 |
Molecular Weight (g/mol) | 74.5kDa |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
16136862
|
Abnova™
H00023029-P01.25UG |
25 ug |
4040.00 DKK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16126862
|
Abnova™
H00023029-P01.10UG |
10 ug |
2665.00 DKK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
Sequence: MSKRKRKRSVQEGENPDDGVRGSPPEDYRLGQVASSLFRGEHHSRGGTGRLASLFSSLEPQIQPVYVPVPKQTIKKTKRNEEEESTSQIERPLSQEPAKKVKAKKKHTNAEKKLADRESALASADLEEEIHQKQGQKRKNSQPGVKVADRKILDDTEDTVVSQRKKIQINQEEERLKNERTVFVGNLPVTCNKKKLKSFFKEYGQIESVRFRSLIPAEGTLSKKLAAIKRKIHPDQKNINAYVVFKEESAATQALKRNGAQIADGFRIRVDLASETSSRDKRSVFVGNLPYKVEESAIEKHFLDCGSIMAVRIVRDKMTGIGKGFGYVLFENTDSVHLALKLNNSELMGRKLRVMRSVNKEKFKQQNSNPRLKNVSKPKQGLNFTSKTAEGHPKSLFIGEKAVLLKTKKKGQKKSGRPKKQRKQKSpecifica
NP_055829.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSKRKRKRSVQEGENPDDGVRGSPPEDYRLGQVASSLFRGEHHSRGGTGRLASLFSSLEPQIQPVYVPVPKQTIKKTKRNEEEESTSQIERPLSQEPAKKVKAKKKHTNAEKKLADRESALASADLEEEIHQKQGQKRKNSQPGVKVADRKILDDTEDTVVSQRKKIQINQEEERLKNERTVFVGNLPVTCNKKKLKSFFKEYGQIESVRFRSLIPAEGTLSKKLAAIKRKIHPDQKNINAYVVFKEESAATQALKRNGAQIADGFRIRVDLASETSSRDKRSVFVGNLPYKVEESAIEKHFLDCGSIMAVRIVRDKMTGIGKGFGYVLFENTDSVHLALKLNNSELMGRKLRVMRSVNKEKFKQQNSNPRLKNVSKPKQGLNFTSKTAEGHPKSLFIGEKAVLLKTKKKGQKKSGRPKKQRKQK | |
RUO | |
RBM34 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23029 | |
RBM34 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0117 | |
RBM34 | |
Recombinant | |
wheat germ expression system |