Learn More
Abnova™ Human RBBP4 Full-length ORF (AAH53904, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00005928-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. It is present in protein complexes involved in histone acetylation and chromatin assembly. It is part of the Mi-2 complex which has been implicated in chromatin remodeling and transcriptional repression associated with histone deacetylation. This encoded protein is also part of co-repressor complexes, which is an integral component of transcriptional silencing. It is found among several cellular proteins that bind directly to retinoblastoma protein to regulate cell proliferation. This protein also seems to be involved in transcriptional repression of E2F-responsive genes. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MADKEAAFDDAVEERVINEEYKIWKKNTPFLYDLVMTHALEWPSLTAQWLPDVTRPEGKDFSIHRLVLGTHTSDEQNHLVIASVQLPNDDAQFDASHYDSEKGEFGGFGSVSGKIEIEIKINHEGEVNRARYMPQNPCIIATKTPSSDVLVFDYTKHPSKPDPSGECNPDLRLRGHQKEGYGLSWNPNLSGHLLSASDDHTICLWDISAVPKEGKVVDAKTIFTGHTAVVEDVSWHLLHESLFGSVADDQKLMIWDTRSNNTSKPSHSVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHSFESHKDEIFQVQWSPHNETILASSGTDRRLNVWDLSKIGEEQSPEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQVWQMAENIYNDEDPEGSVDPEGQGSSpecifications
AAH53904 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.49kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NURF55/RBAP48 | |
RBBP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5928 | |
RBBP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADKEAAFDDAVEERVINEEYKIWKKNTPFLYDLVMTHALEWPSLTAQWLPDVTRPEGKDFSIHRLVLGTHTSDEQNHLVIASVQLPNDDAQFDASHYDSEKGEFGGFGSVSGKIEIEIKINHEGEVNRARYMPQNPCIIATKTPSSDVLVFDYTKHPSKPDPSGECNPDLRLRGHQKEGYGLSWNPNLSGHLLSASDDHTICLWDISAVPKEGKVVDAKTIFTGHTAVVEDVSWHLLHESLFGSVADDQKLMIWDTRSNNTSKPSHSVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHSFESHKDEIFQVQWSPHNETILASSGTDRRLNVWDLSKIGEEQSPEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQVWQMAENIYNDEDPEGSVDPEGQGS | |
RUO | |
RBBP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |