missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBBP4 Full-length ORF (AAH53904, 1 a.a. - 425 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005928-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180102

  • 2665.00 DKK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. It is present in protein complexes involved in histone acetylation and chromatin assembly. It is part of the Mi-2 complex which has been implicated in chromatin remodeling and transcriptional repression associated with histone deacetylation. This encoded protein is also part of co-repressor complexes, which is an integral component of transcriptional silencing. It is found among several cellular proteins that bind directly to retinoblastoma protein to regulate cell proliferation. This protein also seems to be involved in transcriptional repression of E2F-responsive genes. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MADKEAAFDDAVEERVINEEYKIWKKNTPFLYDLVMTHALEWPSLTAQWLPDVTRPEGKDFSIHRLVLGTHTSDEQNHLVIASVQLPNDDAQFDASHYDSEKGEFGGFGSVSGKIEIEIKINHEGEVNRARYMPQNPCIIATKTPSSDVLVFDYTKHPSKPDPSGECNPDLRLRGHQKEGYGLSWNPNLSGHLLSASDDHTICLWDISAVPKEGKVVDAKTIFTGHTAVVEDVSWHLLHESLFGSVADDQKLMIWDTRSNNTSKPSHSVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHSFESHKDEIFQVQWSPHNETILASSGTDRRLNVWDLSKIGEEQSPEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQVWQMAENIYNDEDPEGSVDPEGQGS
Specifications

Specifications

AAH53904
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.49kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NURF55/RBAP48
RBBP4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5928
RBBP4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MADKEAAFDDAVEERVINEEYKIWKKNTPFLYDLVMTHALEWPSLTAQWLPDVTRPEGKDFSIHRLVLGTHTSDEQNHLVIASVQLPNDDAQFDASHYDSEKGEFGGFGSVSGKIEIEIKINHEGEVNRARYMPQNPCIIATKTPSSDVLVFDYTKHPSKPDPSGECNPDLRLRGHQKEGYGLSWNPNLSGHLLSASDDHTICLWDISAVPKEGKVVDAKTIFTGHTAVVEDVSWHLLHESLFGSVADDQKLMIWDTRSNNTSKPSHSVDAHTAEVNCLSFNPYSEFILATGSADKTVALWDLRNLKLKLHSFESHKDEIFQVQWSPHNETILASSGTDRRLNVWDLSKIGEEQSPEDAEDGPPELLFIHGGHTAKISDFSWNPNEPWVICSVSEDNIMQVWQMAENIYNDEDPEGSVDPEGQGS
RUO
RBBP4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers