Få mere at vide
Human RANK/TNFRSF11A (Research Grade Denosumab Biosimilar) Antibody, Novus Biologicals™
GREENER_CHOICE
Human Monoclonal Antibody
Brand:  R&D Systems MAB11492-SP
 Se flere versioner af dette produkt
Produktkode 30114469
  • 1830.00 DKK / 25µg
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
RANK/TNFRSF11A Monoclonal antibody specifically detects RANK/TNFRSF11A in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Tekniske data
Tekniske data
RANK/TNFRSF11A
Monoclonal
Unconjugated
Q9Y6Q6
Human
Protein A or G purified from hybridoma culture supernatant
RUO
8792
Human
Purified
ELISA
Hu200
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFR, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TNFRSF11A, TRANCE R, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Mouse myeloma cell line NS0-derived recombinant human RANK/TNFRSF11A, extracellular domain, Accession # Q9Y6Q6
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG1
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner