missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RANGNRF Full-length ORF (AAI00018.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029098-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MEPTRDCPLFGGAFSAILPMGAIDVSDLRPVPDNQEVFCHPVTDQSLIVELLELQAHVRGEAAARYHFEDVGGVQGARAVHVESVQPLSLENLALRGRCQEAWVLSGKQQIAKENQQVAKDVTLHQALLRLPQYQTDLLLTFNQPPSpecifications
AAI00018.1 | |
Liquid | |
29098 | |
RANGNRF (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEPTRDCPLFGGAFSAILPMGAIDVSDLRPVPDNQEVFCHPVTDQSLIVELLELQAHVRGEAAARYHFEDVGGVQGARAVHVESVQPLSLENLALRGRCQEAWVLSGKQQIAKENQQVAKDVTLHQALLRLPQYQTDLLLTFNQPP | |
RUO | |
RANGRF | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686F02139/HSPC165/HSPC236/MGC110973/MOG1/RANGNRF | |
RANGRF | |
Yes | |
wheat germ expression system |