missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PXN Full-length ORF (XP_001132665.1, 1 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005829-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132058

  • 2665.00 DKK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MPDTFNSRSPSVEGSLWAVGTESQGRDWRHLPTITSELSGAPRCHTVPCAGSTALQEPGEPQGPPASPSCPEEALAATWEQPWASEVFGPERMPPSGAARSFQEVTEPAVVAVDRQAIFPDTWTLTEEHGLQQERPRPEPGRLGSSSPASVTTEQLGAKMTERGSVARPTQGPETPRSPEGTTEAATQDGKEQPELPCAMAMGTPSTTERISTSGQAGTSSHGCWAWPLAGLSVGRSSLTACSLPLCACLLGQKMLRHCGRT
Specifications

Specifications

XP_001132665.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
54.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16691
PXN
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
5829
PXN (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPDTFNSRSPSVEGSLWAVGTESQGRDWRHLPTITSELSGAPRCHTVPCAGSTALQEPGEPQGPPASPSCPEEALAATWEQPWASEVFGPERMPPSGAARSFQEVTEPAVVAVDRQAIFPDTWTLTEEHGLQQERPRPEPGRLGSSSPASVTTEQLGAKMTERGSVARPTQGPETPRSPEGTTEAATQDGKEQPELPCAMAMGTPSTTERISTSGQAGTSSHGCWAWPLAGLSVGRSSLTACSLPLCACLLGQKMLRHCGRT
RUO
PXN
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers