missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Human PSG1 (NP_001171754) VersaClone cDNA

Artikelnummer. 18742788
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18742788 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18742788 Leverandør R&D Systems™ Leverandørnr. RDC2275
  • ONLINE EXCLUSIVE PRICE
    2605.00 DKK
    2460.15 DKK / 10µg
    Spare 144.85 DKK
    5% Slukket
 Begrænset vare
Forventet forsendelse: 14-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

R&D Systems™ Human PSG1 (NP_001171754) VersaClone cDNA is part of the carcinoembryonic antigen (CEA) family and serve as early biochemical markers of syncytiotrophoblast formation.

The Human PSG1 (NP_001171754) VersaClone cDNA clone contains the complete ORF for PSG1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PSG1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Tekniske data

Gene hPSG1
Adgangsnummer NP_001171754
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl
Indsæt type 1273bp
pH 8.5
Gen-id (Entrez) 5669
Gene Alias B1G1, B1G1CD66 antigen-like family member F, CD66f, CD66f antigen, DHFRP2, Fetal liver non-specific cross-reactive antigen 1/2, FLJ90598, FLJ90654, FL-NCA-1/2, PBG1, pregnancy specific beta-1-glycoprotein 1, pregnancy-specific B-1 glycoprotein, Pregnancy-specific beta-1 glycoprotein C/D, pregnancy-specific beta-1-glycoprotein 1, Pregnancy-specific glycoprotein 1, PS-beta-C/D, PS-beta-G-1, PSbG1, PSBG-1, PSBG1SP1, PSG-1, PSG95, PSGGA, PSGGAPSG95, PSGIIA
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Human
Mængde 10 μg
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.