Learn More
Abnova™ Human PRRX2 Full-length ORF (NP_057391.1, 1 a.a. - 253 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051450-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins. Expression is localized to proliferating fetal fibroblasts and the developing dermal layer, with downregulated expression in adult skin. Increases in expression of this gene during fetal but not adult wound healing suggest a possible role in mechanisms that control mammalian dermal regeneration and prevent formation of scar response to wounding. The expression patterns provide evidence consistent with a role in fetal skin development and a possible role in cellular proliferation. [provided by RefSeq]
Sequence: MDSAAAAFALDKPALGPGPPPPPPALGPGDCAQARKNFSVSHLLDLEEVAAAGRLAARPGARAEAREGAAREPSGGSSGSEAAPQDGECPSPGRGSAAKRKKKQRRNRTTFNSSQLQALERVFERTHYPDAFVREELARRVNLSEARVQVWFQNRRAKFRRNERAMLASRSASLLKSYSQEAAIEQPVAPRPTALSPDYLSWTASSPYSTVPPYSPGSSGPATPGVNMANSIASLRLKAKEFSLHHSQVPTVNSpecifications
NP_057391.1 | |
Liquid | |
51450 | |
PRRX2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDSAAAAFALDKPALGPGPPPPPPALGPGDCAQARKNFSVSHLLDLEEVAAAGRLAARPGARAEAREGAAREPSGGSSGSEAAPQDGECPSPGRGSAAKRKKKQRRNRTTFNSSQLQALERVFERTHYPDAFVREELARRVNLSEARVQVWFQNRRAKFRRNERAMLASRSASLLKSYSQEAAIEQPVAPRPTALSPDYLSWTASSPYSTVPPYSPGSSGPATPGVNMANSIASLRLKAKEFSLHHSQVPTVN | |
RUO | |
PRRX2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC19843/PMX2/PRX2 | |
PRRX2 | |
Yes | |
wheat germ expression system |