Få mere at vide
R&D Systems™ Human Proprotein Convertase 9/PCSK9 DuoSet ELISA
Mængde:
1 Kit (for 15 Plates)
Beskrivelse
The Human Proprotein Convertase 9/PCSK9 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Proprotein Convertase 9/PCSK9.
Tekniske data
Tekniske data
| Mængde | 1 Kit (for 15 Plates) |
| Arter | Human |
| Konjugeret | Biotin |
| Analyseområde | 125.00 to 8000 pg/mL |
| Til brug med (applikation) | ELISA |
| Gen-id (Entrez) | 255738 |
| Synonym | EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1 convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 |
| Indhold og opbevaring | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |